Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-1/CD54 Antibody (117G12.02), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. DDX0150A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0150A647 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0150A647 Supplier Novus Biologicals Supplier No. DDX0150A647

Mouse Monoclonal Antibody

ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Sheep samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ICAM-1/CD54
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 117G12.02
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Gene Symbols ICAM1
Host Species Mouse
Immunogen CD54 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3383
Target Species Human, Sheep
Content And Storage Store at -20C in the dark. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.