Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-1/CD54 Antibody (15.2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP250431G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250431G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250431G Supplier Novus Biologicals Supplier No. NBP250431G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen ICAM-1/CD54
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 15.2
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Gene Symbols ICAM1
Host Species Mouse
Immunogen Rheumatoid synovial cells and human monocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3383
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.