Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-1/CD54 Antibody (3E2B) - Azide and BSA Free, Novus Biologicals™

Hamster Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP222540
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Inhibition Activity, CyTOF-ready, Block/Neutralize.Specifications
ICAM-1/CD54 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:10 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Inhibition Activity, CyTOF-ready, Neutralization | |
ICAM1 | |
Mouse ICAM-1 (CD54) | |
500 μg | |
Primary | |
This antibody reacts with Mouse. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Inhibition Assays, CyTOF, Neutralization | |
3E2B | |
Unconjugated | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
Hamster | |
Protein G purified | |
RUO | |
3383 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction