Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICAM-1/CD54 Antibody (3E2B) - Azide and BSA Free, Novus Biologicals™

Hamster Monoclonal Antibody has been used in 1 publication
$196.10 - $501.50
Specifications
Antigen | ICAM-1/CD54 |
---|---|
Clone | 3E2B |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:10 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Inhibition Activity, CyTOF-ready, Neutralization |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Inhibition Assays, CyTOF, Neutralization |
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Inhibition Activity, CyTOF-ready, Block/Neutralize.Specifications
ICAM-1/CD54 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Inhibition Assays, CyTOF, Neutralization | |
Unconjugated | |
Hamster | |
Human, Mouse | |
3383 | |
Mouse ICAM-1 (CD54) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
3E2B | |
Western Blot 1:10 - 1:100, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:200, Inhibition Activity, CyTOF-ready, Neutralization | |
Monoclonal | |
Purified | |
RUO | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
ICAM1 | |
IgG | |
Protein G purified | |
This antibody reacts with Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title