Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-2/CD102 Antibody (015), Alexa Fluor™ 594
SDP

Catalog No. NB066808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB066808 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB066808 Supplier Novus Biologicals Supplier No. NBP289421AF594

Rabbit Monoclonal Antibody

ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Sandwich ELISA Detection.

Specifications

Antigen ICAM-2/CD102
Applications ELISA
Classification Monoclonal
Clone 15
Conjugate Alexa Fluor 594
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (NP_000864.2; Met1-Gln223).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3384
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.