Learn More
Description
Specifications
Specifications
| Antigen | ICAM-2/CD102 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 15 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (NP_000864.2; Met1-Gln223). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
