Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-2/CD102 Antibody (CBRIC2/2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP200320J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200320J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP200320J Supplier Novus Biologicals Supplier No. NBP200320APC

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ICAM-2/CD102
Applications Flow Cytometry
Classification Monoclonal
Clone CBRIC2/2
Conjugate APC
Dilution Flow Cytometry
Gene Alias CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Gene Symbols ICAM2
Host Species Mouse
Immunogen The immunogen for this antibody is ICAM2.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3384
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.