Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-3/CD50 Antibody (186-2G9), HRP, Novus Biologicals™
SDP

Catalog No. NBP234521H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234521H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234521H Supplier Novus Biologicals Supplier No. NBP234521H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ICAM-3/CD50
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 186-2G9
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
Gene Symbols ICAM3
Host Species Mouse
Immunogen Stimulated human leukocytes
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3385
Test Specificity Recognizes the D1 domain of an N-glycosylated glycoprotein of 120kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (Leucocyte Workshop VI). CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This monoclonal antibody is excellent for staining of formalin/paraffin tissues.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.