Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-3/CD50 Antibody (76203), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB373540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373540 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373540 Supplier Novus Biologicals Supplier No. FAB8131E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen ICAM-3/CD50
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 76203
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50, Gln30-His485, Accession # CAA49473
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3385
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.