Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-4 Antibody (729632), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310812 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310812 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310812 Supplier Novus Biologicals Supplier No. FAB8397APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICAM-4 Monoclonal antibody specifically detects ICAM-4 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen ICAM-4
Applications Flow Cytometry
Classification Monoclonal
Clone 729632
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD242, CD242 antigen, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4, Ala23-Ala240, Accession # Q14773
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3386
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.