Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226177 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226177 Supplier Novus Biologicals Supplier No. NBP226177
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ICAM-4 Polyclonal specifically detects ICAM-4 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen ICAM-4
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias CD242, CD242 antigen, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein
Gene Symbols ICAM4
Host Species Goat
Immunogen Peptide with sequence C-RHGSRVIYSESLER corresponding to internal region according to NP_001535.1, NP_071772.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3386
Test Specificity This antibody is expected to recognize isoform 1 (NP_001535.1) and isoform 2 (NP_071772.1) only.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.