Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10053815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10053815 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10053815 Supplier Novus Biologicals Supplier No. NB10053815
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ICAM-5 Polyclonal specifically detects ICAM-5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA.

Specifications

Antigen ICAM-5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_003250.2
Gene Alias ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin
Gene Symbols ICAM5
Host Species Goat
Immunogen Peptide with sequence C-ESPAEGEVFAIQ corresponding to C-Terminus according to NP_003250.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7087
Test Specificity Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Pig, Dog and Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.