Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICBP90 Binding Protein 1 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB185120 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185120 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185120 Supplier Novus Biologicals Supplier No. NBP282094FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ICBP90 Binding Protein 1 Polyclonal antibody specifically detects ICBP90 Binding Protein 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ICBP90 Binding Protein 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias C6orf107, dJ349A12.1, ICBP90, RP3-349A12.1, UHRF1 binding protein 1
Host Species Rabbit
Immunogen ICBP90 Binding Protein 1 antibody was raised against a 17 amino acid synthetic peptide near the center of human ICBP90 Binding Protein 1. The immunogen is located within amino acids 890 - 940 of ICBP90 Binding Protein 1. Amino Acid Squence: DSELSPSEDRELKSDAS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54887
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.