Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICBP90 Binding Protein 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP237702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237702 Supplier Novus Biologicals Supplier No. NBP237702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ICBP90 Binding Protein 1 Polyclonal specifically detects ICBP90 Binding Protein 1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen ICBP90
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060224.3
Gene Alias C6orf107, dJ349A12.1, ICBP90, RP3-349A12.1, UHRF1 binding protein 1
Gene Symbols UHRF1BP1
Host Species Rabbit
Immunogen This antibody maps to a region between residue 1390 to 1440 of human UHRF1 Binding Protein 1 using the numbering given in entry NP_060224.3 (GeneID 54887).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54887
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.