Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ICG-d7

Click to view available options
Quantity:
10 mg
50 mg
Description
ICG-d7 is a partially deuterated form of near-infrared fluorescent dye Indocyanine Green (ICG; Cat. No. 7510). Operates in the 700 - 900 nm wavelength range; also exhibits fluorescence emission in the shortwave infrared window (SWIR; 1,000 - 2,000 nm) which enables in vivo imaging. ICG-d7 shows improved stability in aqueous solutions over ICG with the same fluorescence brightness, and similar absorption and emission maxima in both DMSO and FBS. Excitation maximum = 794 nm; emission maximum = 818 nm.
Specifications
Specifications
Chemical Name or Material | Sodium 4-(2-((1E,3E,5E,7Z)-7-(1,1-Dimethyl-3-(4-sulfonatobutyl)-1,3-dihydro-2H-benzo[e]indol-2-ylidene)hepta-1,3,5-trien-1-yl-1,2,3,4,5,6,7-d7)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate |
CAS | 2861905-57-7 |
Quantity | 10 mg |
Target | Fluorescent Dyes |
Molecular Formula | C43H40D7N2O6S2Na |
Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction