Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ICI 118,551 hydrochloride
GREENER_CHOICE

Catalog No. 082110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-211-0 10 mg
08-215-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 08-211-0 Supplier Tocris Bioscience™ Supplier No. 0821/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective β2 inverse agonist

ICI 118,551 hydrochloride is a very selective β2 adrenergic inverse agonist (Ki values are 1.2, 120 and 257 nM for β2, β1 and β3 receptors respectively). Active in vivo.

Chemical Identifiers

CAS 72795-01-8
Molecular Formula C17H28ClNO2

Specifications

Quantity 10 mg
Formula Weight 313.87
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 313.866
Chemical Name or Material ICI 118,551 hydrochloride
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.