Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Icilin
GREENER_CHOICE

Catalog No. p-4783532
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Activates a novel cold receptor. Cooling agent

Cooling agent that activates the novel cold receptors TRPM8 (CMR1) and TRPA1 (ANKTM1/TRPN1), members of the TRP ion channel family. Induces currents in CMR1-expressing HEK 293 cells (EC50 = 0.36 μM) more potently than menthol or low temperatures. Produces "wet shakes" in vivo.

Chemical Identifiers

CAS 36945-98-9
Molecular Formula C16H13N3O4

Specifications

Quantity 10 mg
Formula Weight 311.3
Purity >98%
Molecular Weight (g/mol) 311.297
Chemical Name or Material Icilin
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.