Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICMT Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB331412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331412 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331412 Supplier Novus Biologicals Supplier No. NBP297509PCP

Rabbit Polyclonal Antibody

ICMT Polyclonal antibody specifically detects ICMT in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen ICMT
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.1.1.100, HSTE14, Isoprenylcysteine Carboxyl Methyltransferase, Isoprenylcysteine carboxylmethyltransferase, MST098, MSTP098, PCCMT, PCMT, PPMT, Prenylated Protein Carboxyl Methyltransferase, Protein-S-Isoprenylcysteine O-Methyltransferase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ICMT fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 23463
Target Species Human, Cynomolgus Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.