Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS Antibody (669214) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB375376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375376 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375376 Supplier Novus Biologicals Supplier No. FAB69753D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction

Specifications

Antigen ICOS
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 669214
Conjugate DyLight 350
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction
Formulation 50mM Sodium Borate
Gene Alias Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombinant human ICOS cocktail, Glu21-Phe141, Accession # Q9Y6W8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 29851
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.