Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS Antibody (669238), Alexa Fluor™ 350, R&D Systems™
SDP

Catalog No. MB69752A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB69752A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB69752A350 Supplier Novus Biologicals Supplier No. MAB69752AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ICOS
Applications Flow Cytometry
Classification Monoclonal
Clone 669238
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Gene Symbols ICOS
Host Species Mouse
Immunogen Partial recombinant human ICOS protein (amino acids 21-141) [UniProt Q9Y6W8]
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 29851.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.