Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS Antibody (669238), DyLight 680, R&D Systems™
SDP

Catalog No. MAB69752FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB69752FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB69752FR Supplier Novus Biologicals Supplier No. MAB69752FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ICOS
Applications Flow Cytometry
Classification Monoclonal
Clone 669238
Conjugate DyLight 680
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Gene Symbols ICOS
Host Species Mouse
Immunogen Partial recombinant human ICOS protein (amino acids 21-141) [UniProt Q9Y6W8]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29851
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.