Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICT Antibody (1E9), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP242633AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242633AG 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242633AG Supplier Novus Biologicals Supplier No. NBP242633AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ICT Monoclonal specifically detects ICT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen ICT
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1000000000
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial
Gene Symbols MRPL58
Host Species Mouse
Immunogen Recombinant human ICT1 (30-206aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3396
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.