Learn More
Description
Specifications
Specifications
| Antigen | ID2 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | 4E12G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389 |
| Gene Symbols | ID2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
