Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB057549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057549 Supplier Novus Biologicals Supplier No. NBP299044F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

ID2 Polyclonal specifically detects ID2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ID2
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ID2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3398
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.