Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227194 0.1 mg
NBP227194SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227194 Supplier Novus Biologicals Supplier No. NBP227194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

ID2 Polyclonal specifically detects ID2 in Bovine, Chicken, Equine, Hamster, Human, Primate, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen ID2
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2ug/ml, Immunocytochemistry/Immunofluorescence reported by customer review
Formulation PBS with 0.05% Sodium Azide
Gene Alias BHLHB26, bHLHb26cell growth-inhibiting gene 8, Class B basic helix-loop-helix protein 26, DNA-binding protein inhibitor ID2, DNA-binding protein inhibitor ID-2, GIG8, helix-loop-helix protein ID2, ID2A, ID2H, inhibitor of differentiation 2, Inhibitor of DNA binding 2, inhibitor of DNA binding 2, dominant negative helix-loop-helix protein, MGC26389
Gene Symbols ID2
Host Species Rabbit
Immunogen A portion of amino acids 60-110 of human ID2 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3398
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, orangutan, rhesus monkey, white-cheeked gibbon, macaque, golden hamster, and mouse, 94% in pig, panda, cow, horse, and rat, 89% in new world monkey, platypus, wild turkey, chicken and Japanese quail, 83% in zebra finch and opossum, and 61% homologous in Carolina anole.
Target Species Bovine, Chicken, Equine, Hamster, Human, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.