Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID3 Antibody (OTI8B3), Novus Biologicals™
SDP

Catalog No. NBP202136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202136 Supplier Novus Biologicals Supplier No. NBP202136
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ID3 Monoclonal specifically detects ID3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ID3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI8B3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50
Gene Alias BHLHB25, bHLHb251R21, Class B basic helix-loop-helix protein 25, DNA-binding protein inhibitor ID-3, HEIR1, HEIR-1, Helix-loop-helix protein HEIR-1, ID-like protein inhibitor HLH 1R21, Inhibitor of DNA binding 3, inhibitor of DNA binding 3, dominant negative helix-loop-helix protein
Gene Symbols ID3
Host Species Mouse
Immunogen Full length human recombinant protein of human ID3 (NP_002158) produced in HEK293T cell.
Molecular Weight of Antigen 12.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3399
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.