Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ID4 Antibody (OTI2G10), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270987R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270987R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270987R Supplier Novus Biologicals Supplier No. NBP270987R

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ID4 Monoclonal specifically detects ID4 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ID4
Applications Western Blot
Classification Monoclonal
Clone OTI2G10
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein
Gene Symbols ID4
Host Species Mouse
Immunogen Full length recombinant protein of human ID4 (NP_001537) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3400.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.