Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Idasanutlin
GREENER_CHOICE

Catalog No. p-7166828
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent MDM2 inhibitor; inhibits MDM2-p53 interaction

Idasanutlin is a potent MDM2 inhibitor (IC50 = 6 nM in binding assay, and 30 nM in cancer cell proliferation assay). Induces p53 stabilisation, cell cycle arrest and apoptosis in cancer cells expressing wildtype p53. Displays inhibition of tumor growth in the SJSA1 tumor xenograft model. Also inhibits MDR-1 at high concentrations.

Specifications

Quantity 10 mg
Molecular Formula C31H29Cl2F2N3O4
Formula Weight 616.49
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.