Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega IdeS Protease

Catalog No. p-6833323 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
5 x 5000 U
5000 U
Catalog No. Quantity
PRV7511 5000 U
PRV7515 5 x 5000 U
2 options

Catalog No. PRV7511

Supplier: Promega V7511

Only null left
Add to Cart
Add to Cart

IdeS Protease is an immunoglobulin-degrading enzyme from Streptococcus pyogenes (IdeS). IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus.

  • Antibody Fragmentation and Characterization, Simplified
  • Highly specific IgG cleavage
  • Rapid generation of highly pure fragments
  • Frozen protease is supplied at a concentration of 50u/μL

Specifications

Content And Storage -30°C to -10°C
Quantity 5000 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.