Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDH2 Antibody (673), Novus Biologicals™
SDP

Catalog No. NBP243572 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243572 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243572 Supplier Novus Biologicals Supplier No. NBP243572
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)

IDH2 Monoclonal specifically detects IDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen IDH2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 673
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS with No Preservative
Gene Alias D2HGA2, EC 1.1.1.42, ICD-M, IDH, IDHM, IDP, IDPM, isocitrate dehydrogenase [NADP], mitochondrial, isocitrate dehydrogenase 2 (NADP+), mitochondrial, mNADP-IDH, NADP(+)-specific ICDH, Oxalosuccinate decarboxylase
Gene Symbols IDH2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human IDH2. The exact sequence is proprietary.
Molecular Weight of Antigen 51 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics, Hypoxia, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3418
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.