Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDH3A Antibody (OTI2F11), PerCP, Novus Biologicals™
SDP

Catalog No. NB270992PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270992PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270992PCP Supplier Novus Biologicals Supplier No. NBP270992PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IDH3A Monoclonal specifically detects IDH3A in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen IDH3A
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2F11
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 1.1.1, EC 1.1.1.41, H-IDH alpha, isocitrate dehydrogenase (NAD+) alpha chain, isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial, isocitrate dehydrogenase 3 (NAD+) alpha, Isocitric dehydrogenase subunit alpha, NAD(+)-specific ICDH subunit alpha, NAD(H)-specific isocitrate dehydrogenase alpha subunit, NAD+-specific ICDH
Gene Symbols IDH3A
Host Species Mouse
Immunogen Full length recombinant protein of human IDH3A (NP_005521) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3419.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.