Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDI2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB112275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112275 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112275 Supplier Novus Biologicals Supplier No. NBP297025AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IDI2 Polyclonal specifically detects IDI2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IDI2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias diphosphate dimethylallyl diphosphate isomerase 2, EC 5.3.3.2, IPP isomerase 2, IPPI2isopentenyl-diphosphate Delta-isomerase 2, Isopentenyl pyrophosphate isomerase 2, isopentenyl-diphosphate delta isomerase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IDI2 (Q9BXS1; Met1-Val227).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91734
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.