Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iduronate 2-Sulfatase/IDS Antibody (OTI1D9), HRP, Novus Biologicals™
SDP

Catalog No. NBP270994H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270994H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270994H Supplier Novus Biologicals Supplier No. NBP270994H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Iduronate 2-Sulfatase/IDS Monoclonal specifically detects Iduronate 2-Sulfatase/IDS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Iduronate 2-Sulfatase/IDS
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D9
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS
Gene Symbols IDS
Host Species Mouse
Immunogen Full length recombinant protein of human Iduronate 2-Sulfatase/IDS (NP_000193) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 3423.0
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.