Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iduronate 2 sulfatase Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A3F9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Iduronate 2 sulfatase |
|---|---|
| Clone | 1A3F9 |
| Concentration | 0.52 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6548
![]()
|
Proteintech Group Inc
661121IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Iduronate-2-sulfatase is required for the lysosomal degradation of heparan sulfate and dermatan sulfate. Mutations in this X-chromosome gene that result in enzymatic deficiency lead to the sex-linked Mucopolysaccharidosis Type II, also known as Hunter Syndrome. Iduronate-2-sulfatase has a strong sequence similarity with human arylsulfatases A, B, and C, and human glucosamine-6-sulfatase. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described.Specifications
| Iduronate 2 sulfatase | |
| 0.52 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| IDS, iduronate 2 sulfatase, Idursulfase | |
| IDS | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1A3F9 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig, Rat | |
| P22304, Q08890 | |
| 100512880, 15931, 3423, 363513 | |
| Iduronate 2 sulfatase Fusion Protein Ag19095 | |
| Primary | |
| -20°C | |
| IDS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title