Learn More
Biocartis Idylla™ Molecular Diagnostics Single-Use Cartridges
Description
- Single-use disposable cartridges
- Polypropylene construction
- Contimaination-controlled design
- All reagents on-board
- Applicable for a wide variety of clinical samples
- All processing steps in one powerful cartridge
- Liquefaction
- Cell lysis
- DNA/RNA extraction
- Real-time PCR amplification and detection
- Data analysis and reporting
- For Research Use Only
Specifications
Specifications
| For Use With (Equipment) | Idylla™ Console and Instruments |
| Quantity | 6 cartridges/box |
| For Use With (Application) | DNA Extraction, Real Time PCR (qPCR) |