Learn More
Biocartis Idylla™ Molecular Diagnostics Single-Use Cartridges
Catalog No. 23555103
Change view
Click to view available options
Quantity:
6 cartridges/box
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-555-103 6 cartridges/box
23-555-100 6 cartridges/box
23-555-101 6 cartridges/box
23-555-102 6 cartridges/box
23-555-104 6 cartridges/box
23-555-105 6 cartridges/box
23-555-106 6 cartridges/box
23-555-110 6 cartridges/box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 23-555-103 Supplier Biocartis Supplier No. A0061/6
Idylla™ Single-Use Cartridges perform all processing steps in one cartridge.
  • Single-use disposable cartridges
  • Polypropylene construction
  • Contimaination-controlled design
  • All reagents on-board
  • Applicable for a wide variety of clinical samples
  • All processing steps in one powerful cartridge
    • Liquefaction
    • Cell lysis
    • DNA/RNA extraction
    • Real-time PCR amplification and detection
    • Data analysis and reporting
    • For Research Use Only
Specifications
For Use With (Equipment) Idylla™ Console and Instruments
Quantity 6 cartridges/box
For Use With (Application) DNA Extraction, Real Time PCR (qPCR)