Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IEM 1460
GSA_VAGREENER_CHOICESDP

Catalog No. 163610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-361-0 10 mg
16-365-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-361-0 Supplier Tocris Bioscience™ Supplier No. 1636/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Open-channel blocker of AMPA currents; selective for non-GluA2-containing receptors

Voltage-dependent open-channel blocker of AMPA receptors. Selective for GluR2 subunit-lacking (Ca2+-permeable) receptors over GluA2-containing receptors (IC50 values are 2.6 and 1102 μM, respectively). Selectively blocks fast spiking interneuron but not medium spiny projection neuron in mouse striatum. Also blocks NMDA receptor-mediated currents. Anticonvulsant in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 121034-89-7
Molecular Formula C19H38Br2N2

Specifications

Quantity 10 mg
Formula Weight 454.33
Purity >98%
Molecular Weight (g/mol) 454.335
Chemical Name or Material IEM 1460
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.