Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IER3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB357359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357359 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357359 Supplier Novus Biologicals Supplier No. NBP176802PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IER3 Polyclonal antibody specifically detects IER3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen IER3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DIF-2, DIF2GLY96, Differentiation-dependent gene 2 protein, expressed in pancreatic carcinoma, gly96, mouse, homolog of, IEX-1, IEX1death protein, IEX-1L, Immediate early protein GLY96, immediate early response 3, Immediate early response 3 protein, immediately early gene X-1, PRG1PACAP-responsive gene 1 protein, Protein DIF-2, Protein PRG1, radiation-inducible immediate-early gene IEX-1
Host Species Rabbit
Immunogen Antibody was raised against a 24 amino acid synthetic peptide near the center of human IEX1. The immunogen is located within amino acids 40 - 90 of IEX-1. Amino Acid Squence: APAGRPSASRGHRKRSRRVLYPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8870
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.