Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IER5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15480920UL
Description
IER5 Polyclonal specifically detects IER5 in Human samples. It is validated for Western Blot.Specifications
IER5 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q5VY09 | |
IER5 | |
Synthetic peptides corresponding to IER5(immediate early response 5) The peptide sequence was selected from the N terminal of IER5. Peptide sequence MEFKLEAHRIVSISLGKIYNSRVQRGGIKLHKNLLVSLVLRSARQVYLSD. | |
Affinity Purified | |
RUO | |
51278 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
immediate early response 5, immediate early response gene 5 protein, MGC102760, SBBI48 | |
Rabbit | |
34 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction