Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IER5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | IER5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15480920
![]() |
Novus Biologicals
NBP15480920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154809
![]() |
Novus Biologicals
NBP154809 |
100 μL |
Each for $487.50
|
|
|||||
Description
IER5 Polyclonal specifically detects IER5 in Human samples. It is validated for Western Blot.Specifications
IER5 | |
Polyclonal | |
Rabbit | |
Q5VY09 | |
51278 | |
Synthetic peptides corresponding to IER5(immediate early response 5) The peptide sequence was selected from the N terminal of IER5. Peptide sequence MEFKLEAHRIVSISLGKIYNSRVQRGGIKLHKNLLVSLVLRSARQVYLSD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
immediate early response 5, immediate early response gene 5 protein, MGC102760, SBBI48 | |
IER5 | |
IgG | |
34 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title