Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IER5L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | IER5L |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17961020
![]() |
Novus Biologicals
NBP17961020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179610
![]() |
Novus Biologicals
NBP179610 |
100 μL |
Each for $487.50
|
|
|||||
Description
IER5L Polyclonal specifically detects IER5L in Human samples. It is validated for Western Blot.Specifications
IER5L | |
Polyclonal | |
Rabbit | |
NP_982258 | |
389792 | |
Synthetic peptide directed towards the middle region of human IER5LThe immunogen for this antibody is IER5L. Peptide sequence SNLISIFGSGFSGLVSRQPDSSEQPPPLNGQLCAKQALASLGAWTRAIVA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
bA247A12.2, immediate early response 5-like, immediate early response gene 5-like protein, MGC70833 | |
IER5L | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title