Learn More
Description
Specifications
Specifications
| Antigen | IF3EI |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q9Y262 |
| Gene Alias | eIEF associated protein HSPC021, EIF3EIP, eIF3l, EIF3S11, EIF3S6IP, Eukaryotic translation initiation factor 3 subunit 6-interacting protein, Eukaryotic translation initiation factor 3 subunit E-interacting protein, eukaryotic translation initiation factor 3 subunit L, eukaryotic translation initiation factor 3, subunit 6 interacting protein, eukaryotic translation initiation factor 3, subunit L, HSPC021, HSPC025, MSTP005, subunit E interacting protein |
| Gene Symbols | EIF3L |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
