Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IF3EI Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen IF3EI
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156926
SDP
View Documents
Novus Biologicals
NBP156926
100 μL
Each for $480.74
Only null left
Add to Cart
 
NBP15692620
SDP
View Documents
Novus Biologicals
NBP15692620UL
20 μL N/A N/A N/A
Description

Description

IF3EI Polyclonal specifically detects IF3EI in Human samples. It is validated for Western Blot.
Specifications

Specifications

IF3EI
Polyclonal
Rabbit
Core ESC Like Genes, Stem Cell Markers
eIEF associated protein HSPC021, EIF3EIP, eIF3l, EIF3S11, EIF3S6IP, Eukaryotic translation initiation factor 3 subunit 6-interacting protein, Eukaryotic translation initiation factor 3 subunit E-interacting protein, eukaryotic translation initiation factor 3 subunit L, eukaryotic translation initiation factor 3, subunit 6 interacting protein, eukaryotic translation initiation factor 3, subunit L, HSPC021, HSPC025, MSTP005, subunit E interacting protein
EIF3L
Primary
Western Blot
Unconjugated
RUO
Q9Y262
51386
IgG
This product is specific to Subunit or Isoform: L.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.