Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI202b Antibody, Novus Biologicals™
SDP

Catalog No. NBP227066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227066 0.1 mg
NBP227066SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227066 Supplier Novus Biologicals Supplier No. NBP227066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

IFI202b Polyclonal specifically detects IFI202b in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen IFI202b
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9R002
Gene Symbols IFI202B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 273-290 of the mouse Ifi202b protein.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26388
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.