Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI27 Antibody, Novus Biologicals™
SDP

Catalog No. NB300709005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300709005 0.05 mg
NB018174 0.025 mg
NB300709 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300709005 Supplier Novus Biologicals Supplier No. NB3007090.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFI27 Polyclonal specifically detects IFI27 in Human, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen IFI27
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:10000-1:1000
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27
Gene Symbols IFI27
Host Species Rabbit
Immunogen Synthetic peptide.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 3429
Test Specificity IFI27
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.