Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFI44 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$158.00 - $499.50

Specifications

Antigen IFI44
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15538020
SDP
View Documents
Novus Biologicals
NBP15538020UL
20 μL
Each for $158.00
Only null left
Add to Cart
 
NBP155380
SDP
View Documents
Novus Biologicals
NBP155380
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

IFI44 Polyclonal specifically detects IFI44 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IFI44
Polyclonal
Rabbit
Human
interferon-induced protein 44, interferon-induced, hepatitis C-associated microtubular aggregate protein(44kD), MTAP44p44Microtubule-associated protein 44, P44
IFI44
IgG
50 kDa
Western Blot
Unconjugated
RUO
Q8TCB0
10561
Synthetic peptides corresponding to IFI44(interferon-induced protein 44) The peptide sequence was selected from the middle region of IFI44. Peptide sequence LIEIERCEPVRSKLEEVQRKLGFALSDISVVSNYSSEWELDPVKDVLILS.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.