Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFIT1 Mouse anti-Mouse, Unconjugated, Clone: 13B4-1, MilliporeSigma™
Mouse Monoclonal Antibody
$184.08 - $461.76
Specifications
Antigen | IFIT1 |
---|---|
Clone | 13B4-1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF22282MI
|
MilliporeSigma
MABF222825UG |
25 μg |
Each for $184.08
|
|
|||||
MABF22281MI
|
MilliporeSigma
MABF2228100UG |
100 μg |
Each for $461.76
|
|
|||||
Description
IFIT1 Monoclonal antibody specifically detects IFIT1 in Mouse samples. It is validated for Western Blotting.Specifications
IFIT1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse | |
Interferon-induced protein with tetratricopeptide repeats 1;IFIT-1;Glucocorticoid-attenuated response gene 16 protein;GARG-16;Interferon-induced 56 kDa protein;IFI-56K;P56 | |
His-tagged full-length recombinant mouse Interferon-induced protein with tetratricopeptide repeats 1 (ITIT1). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼53 kDa observed; 53.74 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
13B4-1 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
NP_032357 | |
Ifit1, Garg16, Ifi56, Isg56 | |
IgG1 κ | |
Protein G Purified | |
Clone 13B4-1 specifically detects murine Interferon-induced protein with tetratricopeptide repeats 1 (IFIT1). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title