Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFIT2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215180SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215180SS 0.025 mg
NBP215180 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP215180SS Supplier Novus Biologicals Supplier No. NBP215180SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

IFIT2 Polyclonal specifically detects IFIT2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IFIT2
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_001538.4
Gene Alias cig42, G10P2IFI-54K, GARG-39, IFI-54, IFI54IFIT-2, IFI-54K, Interferon, alpha-inducible protein (MW 54kD), Interferon-induced 54 kDa protein, interferon-induced protein 54, interferon-induced protein with tetratricopeptide repeats 2, ISG54, ISG-54 K, ISG-54K, P54
Gene Symbols IFIT2
Host Species Rabbit
Immunogen A portion of amino acids 320-370 from human IFIT2 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3433
Test Specificity Reacts with Human and Mouse. The IFIT2 amino acid sequence used as immunogen for this antibody is 100% homologous in human, 94% in new world monkey, rhesus monkey, white-cheeked gibbon, chimpanzee, and orangutan, 72% in cow, dog, Chinese hamster, mole rat, and rabbit, 66% in mouse, and 61% homologouus in pig.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.