Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFITM1 Antibody (001), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB088035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088035 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088035 Supplier Novus Biologicals Supplier No. NBP289229AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IFITM1 Monoclonal specifically detects IFITM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IFITM1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias CD225 antigen, CD225Leu-13 antigen, IFI17LEU13,9-27, interferon induced transmembrane protein 1 (9-27), Interferon-induced protein 17, interferon-induced transmembrane protein 1, Interferon-inducible protein 9-27
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human IFITM1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8519
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.