Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFITM3/Fragilis Antibody, Novus Biologicals™
SDP

Catalog No. NB6001534 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001534 0.1 mg
NB6001534SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001534 Supplier Novus Biologicals Supplier No. NB6001534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFITM3/Fragilis Polyclonal specifically detects IFITM3/Fragilis in Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen IFITM3/Fragilis
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Simple Western 1:10
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9CQW9
Gene Alias 1-8U, interferon induced transmembrane protein 3 (1-8U), interferon-induced transmembrane protein 3, Interferon-inducible protein 1-8U, IP15
Gene Symbols IFITM3
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of mouse fragilis was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10410
Test Specificity Fragilis - Germ Line Marker
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.